Product Details
- SNP ID
-
hCV31373202
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:95488477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCCTAGCTGCTAGTAGATCTAAG[G/T]GGTTATATCAATTTCTTAAGTCATA
- Phenotype
-
MIM: 605761
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ASB4
PubMed Links
Gene Details
- Gene
- ASB4
- Gene Name
- ankyrin repeat and SOCS box containing 4
There are no transcripts associated with this gene.
View Full Product Details