Product Details

SNP ID
rs12814229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15622852 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAGGCTCTTACTCACTCAGAAG[A/C]TAATTTCCATTGTTATGTCAAAATT
Phenotype
MIM: 600206
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EPS8 PubMed Links

Gene Details

Gene
EPS8
Gene Name
epidermal growth factor receptor pathway substrate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004447.5 Intron NP_004438.3
XM_011520605.2 Intron XP_011518907.1

View Full Product Details