Product Details

SNP ID
rs4931412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27770021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGATTCCATGTGACAGACAAAAC[A/G]TCTATGAGGAATATTTGGAAATATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL42 PubMed Links

Gene Details

Gene
KLHL42
Gene Name
kelch like family member 42
There are no transcripts associated with this gene.

Gene
MANSC4
Gene Name
MANSC domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146221.1 Intron NP_001139693.1
XM_006719024.3 Intron XP_006719087.1
XM_011520542.2 Intron XP_011518844.1

View Full Product Details