Product Details

SNP ID
rs12326772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50797223 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGGACAACTTGTGTCATGTGTCC[A/G]GGCCTCAGACTCTGCTGTGGGTTGG
Phenotype
MIM: 608080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRO PubMed Links

Gene Details

Gene
MRO
Gene Name
maestro
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127174.1 2932 UTR 3 NP_001120646.1
NM_001127175.1 2932 UTR 3 NP_001120647.1
NM_001127176.1 2932 UTR 3 NP_001120648.1
NM_031939.3 2932 UTR 3 NP_114145.2
XM_005258365.3 2932 UTR 3 XP_005258422.1
XM_011526226.2 2932 UTR 3 XP_011524528.1
XM_017026035.1 2932 UTR 3 XP_016881524.1
XM_017026036.1 2932 UTR 3 XP_016881525.1
XM_017026037.1 2932 UTR 3 XP_016881526.1

View Full Product Details