Product Details

SNP ID
rs6677422
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:100998679 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTTATTTAGTGCCTACACTGTG[C/T]TAGACTTCTAGACATTAGGAAGAGA
Phenotype
MIM: 611075 MIM: 611149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPH5 PubMed Links

Gene Details

Gene
DPH5
Gene Name
diphthamide biosynthesis 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077394.1 Intron NP_001070862.1
NM_001077395.1 Intron NP_001070863.1
NM_015958.2 Intron NP_057042.2
XM_005270938.1 Intron XP_005270995.1
XM_005270939.4 Intron XP_005270996.1
XM_005270941.3 Intron XP_005270998.1
XM_006710683.3 Intron XP_006710746.1
XM_011541569.1 Intron XP_011539871.1
XM_011541570.2 Intron XP_011539872.1
XM_017001460.1 Intron XP_016856949.1
XM_017001461.1 Intron XP_016856950.1
XM_017001462.1 Intron XP_016856951.1
Gene
SLC30A7
Gene Name
solute carrier family 30 member 7
There are no transcripts associated with this gene.

View Full Product Details