Product Details
- SNP ID
-
rs11591183
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:95096150 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTTCAAGCAATCTGAGGTTATTC[C/T]CTACCAATTAGCCATTCAAATTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM56
PubMed Links
Gene Details
- Gene
- TMEM56
- Gene Name
- transmembrane protein 56
View Full Product Details