Product Details

SNP ID
rs11804508
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:228211781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTTGCAGGTCCCCGCCGCCACC[C/G]TCATGGATCAGCCACAGTTCAGCGG
Phenotype
MIM: 608616
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf145 PubMed Links

Gene Details

Gene
C1orf145
Gene Name
chromosome 1 open reading frame 145
There are no transcripts associated with this gene.

Gene
OBSCN
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098623.2 151 UTR 5 NP_001092093.2
NM_001271223.2 151 UTR 5 NP_001258152.2
NM_052843.3 151 UTR 5 NP_443075.3
XM_005273287.4 151 UTR 5 XP_005273344.1
XM_005273291.4 151 UTR 5 XP_005273348.1
XM_005273298.4 151 UTR 5 XP_005273355.1
XM_005273307.4 151 UTR 5 XP_005273364.1
XM_006711822.3 151 UTR 5 XP_006711885.1
XM_006711823.3 151 UTR 5 XP_006711886.1
XM_006711827.3 151 UTR 5 XP_006711890.1
XM_006711829.3 151 UTR 5 XP_006711892.1
XM_011544297.2 151 UTR 5 XP_011542599.1
XM_011544299.2 151 UTR 5 XP_011542601.1
XM_017002443.1 151 UTR 5 XP_016857932.1
XM_017002444.1 151 UTR 5 XP_016857933.1
XM_017002445.1 151 UTR 5 XP_016857934.1
XM_017002446.1 151 UTR 5 XP_016857935.1
XM_017002447.1 151 UTR 5 XP_016857936.1
XM_017002448.1 151 UTR 5 XP_016857937.1
XM_017002449.1 151 UTR 5 XP_016857938.1
XM_017002450.1 151 UTR 5 XP_016857939.1
XM_017002451.1 151 UTR 5 XP_016857940.1
XM_017002452.1 151 UTR 5 XP_016857941.1
XM_017002453.1 151 UTR 5 XP_016857942.1
XM_017002454.1 151 UTR 5 XP_016857943.1
XM_017002455.1 151 UTR 5 XP_016857944.1
XM_017002456.1 151 UTR 5 XP_016857945.1
XM_017002457.1 151 UTR 5 XP_016857946.1
XM_017002458.1 151 UTR 5 XP_016857947.1
XM_017002459.1 151 UTR 5 XP_016857948.1
XM_017002460.1 151 UTR 5 XP_016857949.1
XM_017002461.1 151 UTR 5 XP_016857950.1
XM_017002462.1 151 UTR 5 XP_016857951.1
XM_017002463.1 151 UTR 5 XP_016857952.1
XM_017002464.1 151 UTR 5 XP_016857953.1
XM_017002465.1 151 UTR 5 XP_016857954.1
XM_017002466.1 151 UTR 5 XP_016857955.1
XM_017002467.1 151 UTR 5 XP_016857956.1
XM_017002468.1 151 UTR 5 XP_016857957.1
XM_017002469.1 151 UTR 5 XP_016857958.1
XM_017002470.1 151 UTR 5 XP_016857959.1
XM_017002471.1 151 UTR 5 XP_016857960.1
XM_017002472.1 151 UTR 5 XP_016857961.1
XM_017002473.1 151 UTR 5 XP_016857962.1

View Full Product Details