Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170637.3 | Intron | NP_001164108.1 | |||
NM_001300952.1 | Intron | NP_001287881.1 | |||
NM_015326.4 | Intron | NP_056141.2 | |||
XM_005277510.2 | Intron | XP_005277567.1 | |||
XM_005277511.2 | Intron | XP_005277568.1 | |||
XM_005277514.1 | Intron | XP_005277571.1 | |||
XM_005277515.1 | Intron | XP_005277572.1 | |||
XM_011509354.1 | Intron | XP_011507656.1 | |||
XM_011509355.1 | Intron | XP_011507657.1 | |||
XM_011509356.2 | Intron | XP_011507658.1 | |||
XM_011509359.1 | Intron | XP_011507661.1 | |||
XM_017000838.1 | Intron | XP_016856327.1 | |||
XM_017000839.1 | Intron | XP_016856328.1 | |||
XM_017000840.1 | Intron | XP_016856329.1 | |||
XM_017000841.1 | Intron | XP_016856330.1 |