Product Details

SNP ID
rs12611158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12075457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCAGTGTGTGTGGAGAAGTCTTC[A/G]TGGGTCATTCTTCCCTTAATAGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF844 PubMed Links

Gene Details

Gene
ZNF844
Gene Name
zinc finger protein 844
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136501.2 358 Missense Mutation ATG,GTG M113V NP_001129973.1
XM_017026664.1 358 Missense Mutation ATG,GTG M115V XP_016882153.1

View Full Product Details