Product Details

SNP ID
rs4804737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12837124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTGTCATTATAAAAAATGACTA[C/T]GCGGCCATCACCTGAATTTCCATTT
Phenotype
MIM: 612256 MIM: 609553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST1 PubMed Links

Gene Details

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014975.2 Intron NP_055790.1
XM_011527805.2 Intron XP_011526107.1
XM_011527808.2 Intron XP_011526110.1
XM_017026501.1 Intron XP_016881990.1
Gene
RTBDN
Gene Name
retbindin
There are no transcripts associated with this gene.

View Full Product Details