Product Details

SNP ID
rs12042999
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:156768636 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCTCCTGAACCTTTGCCGAACA[C/T]CTGGGACAAATCTTTTCCTTGCCTC
Phenotype
MIM: 179755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRCC PubMed Links

Gene Details

Gene
PRCC
Gene Name
papillary renal cell carcinoma (translocation-associated)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005973.4 Intron NP_005964.3
XM_005245313.2 Intron XP_005245370.1
XM_005245314.1 Intron XP_005245371.1
XM_011509726.1 Intron XP_011508028.1

View Full Product Details