Product Details

SNP ID
rs11804423
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109219880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATTAACATGTAGCAGTAGTTGG[G/T]TCTTTTCATTGCTATATAATACTCC
Phenotype
MIM: 607529
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SARS PubMed Links

Gene Details

Gene
SARS
Gene Name
seryl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006513.3 Intron NP_006504.2
XM_006710813.2 Intron XP_006710876.1

View Full Product Details