Product Details
- SNP ID
-
rs7178777
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:41342390 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAATAAGGTCTCTCTTGCAGGCAA[A/C]CAAGTTGTTAAAAGCCTTGAAAGGC
- Phenotype
-
MIM: 612818
MIM: 606020
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NUSAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7178634] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUSAP1
- Gene Name
- nucleolar and spindle associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243142.1 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
NP_001230071.1 |
NM_001243143.1 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
NP_001230072.1 |
NM_001243144.1 |
216 |
Intron |
|
|
NP_001230073.1 |
NM_001301136.1 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
NP_001288065.1 |
NM_016359.4 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
NP_057443.2 |
NM_018454.7 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
NP_060924.4 |
XM_005254428.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_005254485.1 |
XM_005254430.4 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_005254487.1 |
XM_005254431.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_005254488.1 |
XM_006720559.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_006720622.1 |
XM_006720560.3 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_006720623.1 |
XM_006720561.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_006720624.1 |
XM_006720562.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_006720625.1 |
XM_006720563.2 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_006720626.1 |
XM_017022294.1 |
216 |
Missense Mutation |
AAC,ACC |
N33T |
XP_016877783.1 |
XM_017022295.1 |
216 |
UTR 5 |
|
|
XP_016877784.1 |
- Gene
- OIP5
- Gene Name
- Opa interacting protein 5
There are no transcripts associated with this gene.
View Full Product Details