Product Details

SNP ID
rs12858309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71284114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCTCCTCCTCTCGGCCCCGCG[A/T]GGCCATTAATTCAGAATGGGCAAGT
Phenotype
MIM: 300084
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NONO PubMed Links

Gene Details

Gene
NONO
Gene Name
non-POU domain containing, octamer-binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145408.1 Intron NP_001138880.1
NM_001145409.1 Intron NP_001138881.1
NM_001145410.1 Intron NP_001138882.1
NM_007363.4 Intron NP_031389.3

View Full Product Details