Product Details

SNP ID
rs11667949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51715673 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTATCCCAAGTTTCCTCTGGGT[A/C]TTTCAGTGGACCTGGCTTTGCAGAA
Phenotype
MIM: 601463
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAS1 PubMed Links

Gene Details

Gene
HAS1
Gene Name
hyaluronan synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297436.1 Intron NP_001284365.1
NM_001523.3 Intron NP_001514.2
XM_011526884.2 Intron XP_011525186.1
Gene
SPACA6
Gene Name
sperm acrosome associated 6
There are no transcripts associated with this gene.

View Full Product Details