Product Details

SNP ID
rs3760905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4182942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCTAAGGGTTTTGAATGCCAC[G/T]CAGAGTTACCTGAATATCTCATTTG
Phenotype
MIM: 611998 MIM: 606211
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD24 PubMed Links

Gene Details

Gene
ANKRD24
Gene Name
ankyrin repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133475.1 Intron NP_597732.1
XM_011527756.2 Intron XP_011526058.1
XM_011527757.2 Intron XP_011526059.1
XM_011527758.2 Intron XP_011526060.1
XM_011527761.1 Intron XP_011526063.1
XM_011527764.2 Intron XP_011526066.1
XM_011527765.2 Intron XP_011526067.1
XM_011527768.2 Intron XP_011526070.1
XM_017026438.1 Intron XP_016881927.1
XM_017026439.1 Intron XP_016881928.1
XM_017026440.1 Intron XP_016881929.1
XM_017026441.1 Intron XP_016881930.1
Gene
CREB3L3
Gene Name
cAMP responsive element binding protein 3 like 3
There are no transcripts associated with this gene.

Gene
SIRT6
Gene Name
sirtuin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193285.2 Intron NP_001180214.1
NM_001321058.1 Intron NP_001307987.1
NM_001321059.1 Intron NP_001307988.1
NM_001321060.1 Intron NP_001307989.1
NM_001321061.1 Intron NP_001307990.1
NM_001321062.1 Intron NP_001307991.1
NM_001321063.1 Intron NP_001307992.1
NM_001321064.1 Intron NP_001307993.1
NM_016539.3 Intron NP_057623.2

View Full Product Details