Product Details
- SNP ID
-
rs12534948
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:66936470 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCGGTTTGCCAGTATGTTTTGGA[G/A]GAGATTTACAACAATCTTCATCAGG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM248
PubMed Links
Gene Details
- Gene
- TMEM248
- Gene Name
- transmembrane protein 248
View Full Product Details