Product Details

SNP ID
rs3918164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150994085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCAGGTCACAAATGCAAAAGGG[C/T]TATTAATGTGCATAGAACAGGACAG
Phenotype
MIM: 163729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOS3 PubMed Links

Gene Details

Gene
NOS3
Gene Name
nitric oxide synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000603.4 Intron NP_000594.2
NM_001160109.1 Intron NP_001153581.1
NM_001160110.1 Intron NP_001153582.1
NM_001160111.1 Intron NP_001153583.1
XM_006716002.3 Intron XP_006716065.1
XM_017012232.1 Intron XP_016867721.1
XM_017012233.1 Intron XP_016867722.1
XM_017012234.1 Intron XP_016867723.1

View Full Product Details