Product Details

SNP ID
rs8191494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88810108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATCATCCACGACGACCACCCTC[T/C]GTCCTGGCTCCAGGGCGTCTTTCTG
Phenotype
MIM: 102600 MIM: 605525 MIM: 612222
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
APRT PubMed Links

Gene Details

Gene
APRT
Gene Name
adenine phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000485.2 397 Missense Mutation CAG,CGG Q121R NP_000476.1
NM_001030018.1 397 Missense Mutation CAG,CGG Q121R NP_001025189.1
Gene
CDT1
Gene Name
chromatin licensing and DNA replication factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030928.3 397 Intron NP_112190.2
Gene
GALNS
Gene Name
galactosamine (N-acetyl)-6-sulfatase
There are no transcripts associated with this gene.

View Full Product Details