Product Details

SNP ID
rs12672765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73447124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACCCATTTAAACCCATGGCAG[C/T]TAAGTTACGCCACAGTCACAACACA
Phenotype
MIM: 605681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAZ1B PubMed Links

Gene Details

Gene
BAZ1B
Gene Name
bromodomain adjacent to zinc finger domain 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032408.3 Intron NP_115784.1
XM_017012773.1 Intron XP_016868262.1

View Full Product Details