Product Details

SNP ID
rs12815700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132997752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTAAAACAATGAGAGATACATA[A/C]CAATAATTTGAATAATAGAAATAGA
Phenotype
MIM: 194537
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF26 PubMed Links

Gene Details

Gene
ZNF26
Gene Name
zinc finger protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256279.1 Intron NP_001243208.1
NM_001256280.1 Intron NP_001243209.1
NM_019591.3 Intron NP_062537.2
XM_005266182.3 Intron XP_005266239.1
XM_005266183.3 Intron XP_005266240.1
XM_011534829.1 Intron XP_011533131.1
XM_017019920.1 Intron XP_016875409.1
XM_017019921.1 Intron XP_016875410.1
XM_017019922.1 Intron XP_016875411.1
XM_017019923.1 Intron XP_016875412.1

View Full Product Details