Product Details

SNP ID
rs4732416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75812444 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAAAATGAGTTCACAGTTACTAC[C/T]TATGAAGCTATTTGGAGGACAGACA
Phenotype
MIM: 602495
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL24 PubMed Links

Gene Details

Gene
CCL24
Gene Name
C-C motif chemokine ligand 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002991.2 Intron NP_002982.2
XM_011516459.2 Intron XP_011514761.1
XM_011516460.2 Intron XP_011514762.1

View Full Product Details