Product Details

SNP ID
rs12067522
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:150553409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTCAGAGCTGTGCCCCCACAT[C/T]TGAAACACCTTCTCAGGTCCCGGCT
Phenotype
MIM: 610113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL4 PubMed Links

Gene Details

Gene
ADAMTSL4
Gene Name
ADAMTS like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288607.1 Intron NP_001275536.1
NM_001288608.1 Intron NP_001275537.1
NM_019032.5 Intron NP_061905.2
NM_025008.4 Intron NP_079284.2
XM_011509644.2 Intron XP_011507946.1
XM_011509645.2 Intron XP_011507947.1
XM_011509648.2 Intron XP_011507950.1
XM_011509649.2 Intron XP_011507951.1
XM_011509650.2 Intron XP_011507952.1
XM_011509651.2 Intron XP_011507953.1
XM_011509652.1 Intron XP_011507954.1
XM_017001506.1 Intron XP_016856995.1
XM_017001507.1 Intron XP_016856996.1
Gene
ADAMTSL4-AS1
Gene Name
ADAMTSL4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC100289061
Gene Name
uncharacterized LOC100289061
There are no transcripts associated with this gene.

Gene
LOC107983947
Gene Name
uncharacterized LOC107983947
There are no transcripts associated with this gene.

Gene
MIR4257
Gene Name
microRNA 4257
There are no transcripts associated with this gene.

View Full Product Details