Product Details

SNP ID
rs11205357
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:150295378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCTCCTGACTAGCTGGGTCTAG[C/T]GACGCACGCCACCACCGCAGTTAAT
Phenotype
MIM: 615782 MIM: 611984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIART PubMed Links

Gene Details

Gene
CIART
Gene Name
circadian associated repressor of transcription
There are no transcripts associated with this gene.

Gene
MRPS21
Gene Name
mitochondrial ribosomal protein S21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018997.3 Intron NP_061870.1
NM_031901.5 Intron NP_114107.1

View Full Product Details