Product Details

SNP ID
rs11585929
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:150273123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTAGCAACACTGAGCTCAGTT[C/T]TCCATCACCACGGAAGTTGAACAAG
Phenotype
MIM: 607629 MIM: 604832 MIM: 615782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077628.2 Intron NP_001071096.1
NM_001243771.1 Intron NP_001230700.1
NM_001243772.1 Intron NP_001230701.1
NM_016022.3 Intron NP_057106.2
XM_017001417.1 Intron XP_016856906.1
Gene
C1orf54
Gene Name
chromosome 1 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301039.1 Intron NP_001287968.1
NM_001301040.1 Intron NP_001287969.1
NM_001301041.1 Intron NP_001287970.1
NM_001301042.1 Intron NP_001287971.1
NM_024579.3 Intron NP_078855.2
XM_011509984.2 Intron XP_011508286.1
Gene
CA14
Gene Name
carbonic anhydrase 14
There are no transcripts associated with this gene.

Gene
CIART
Gene Name
circadian associated repressor of transcription
There are no transcripts associated with this gene.

View Full Product Details