Product Details

SNP ID
rs12120851
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:114720224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACGTGGTTTCTGTGTTTGTGTGA[C/T]ATGCAAAAGAAGCTCTAAGCAAAAG
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007553.2 Intron NP_001007554.1
NM_001130523.2 Intron NP_001123995.1
NM_001242891.1 Intron NP_001229820.1
NM_001242892.1 Intron NP_001229821.1
NM_001242893.1 Intron NP_001229822.1
NM_007158.5 Intron NP_009089.4
Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
There are no transcripts associated with this gene.

View Full Product Details