Product Details

SNP ID
rs7853065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34204243 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGGGAGGCTAAGGCACAAGAATCT[C/T]GCTGCTGCACTCCAGAGTGCAGACG
Phenotype
MIM: 609787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBAP1 PubMed Links

Gene Details

Gene
UBAP1
Gene Name
ubiquitin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171201.1 Intron NP_001164672.1
NM_001171202.1 Intron NP_001164673.1
NM_001171203.2 Intron NP_001164674.1
NM_001171204.2 Intron NP_001164675.1
NM_016525.4 Intron NP_057609.2
XM_006716779.3 Intron XP_006716842.1
XM_011517898.2 Intron XP_011516200.1
XM_011517899.1 Intron XP_011516201.1
XM_011517900.1 Intron XP_011516202.1
XM_017014801.1 Intron XP_016870290.1
XM_017014802.1 Intron XP_016870291.1

View Full Product Details