Product Details

SNP ID
rs3138158
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:6460252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTGGGAGCCGCACAGCCCTCCC[C/T]TCCTGCGCTGGAAACCCTCCCTGAC
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031475.2 2839 UTR 3 NP_113663.2
XM_005263501.2 2839 UTR 3 XP_005263558.1
XM_011542231.1 2839 Intron XP_011540533.1
XM_011542232.1 2839 Intron XP_011540534.1
XM_011542233.2 2839 Intron XP_011540535.1
XM_011542236.2 2839 Intron XP_011540538.1
XM_011542237.1 2839 Intron XP_011540539.1
XM_011542238.2 2839 Intron XP_011540540.1
XM_017002433.1 2839 Intron XP_016857922.1
XM_017002434.1 2839 Intron XP_016857923.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039664.1 2839 Intron NP_001034753.1
NM_003790.2 2839 Intron NP_003781.1
NM_148965.1 2839 Intron NP_683866.1
NM_148966.1 2839 Intron NP_683867.1
NM_148967.1 2839 Intron NP_683868.1
NM_148970.1 2839 Intron NP_683871.1

View Full Product Details