Product Details

SNP ID
rs4801939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52848448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTCTGAATGCTGAGGGAGCACT[A/G]TAATATGTAATATGTGGACATCAAG
Phenotype
MIM: 616841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF468 PubMed Links

Gene Details

Gene
ZNF468
Gene Name
zinc finger protein 468
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008801.1 Intron NP_001008801.1
NM_001277120.1 Intron NP_001264049.1
XM_017027443.1 Intron XP_016882932.1
XM_017027444.1 Intron XP_016882933.1

View Full Product Details