Product Details

SNP ID
rs12323452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:66490324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGAATCAGAAAGATCTGGTTC[A/C]AATTCCAACTCTGTACTTTCCTAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00238 PubMed Links

Gene Details

Gene
LINC00238
Gene Name
long intergenic non-protein coding RNA 238
There are no transcripts associated with this gene.

View Full Product Details