Product Details

SNP ID
rs6943066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141934877 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCAAGGAAAAGGTACATGGATG[G/T]GTTTTGGTCAAGCAAATTGGGATCC
Phenotype
MIM: 604987
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC5A PubMed Links

Gene Details

Gene
CLEC5A
Gene Name
C-type lectin domain family 5 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301167.1 Intron NP_001288096.1
NM_013252.2 Intron NP_037384.1
XM_011515995.2 Intron XP_011514297.1
XM_017011915.1 Intron XP_016867404.1
XM_017011916.1 Intron XP_016867405.1
XM_017011917.1 Intron XP_016867406.1

View Full Product Details