Product Details

SNP ID
rs11835821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1949127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCTCCTGGAGCTGGAGCTTGG[A/T]GAGTGGGCTGCTGGTGATCACGGAG
Phenotype
MIM: 609843
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCP1B PubMed Links

Gene Details

Gene
DCP1B
Gene Name
decapping mRNA 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319292.1 1957 Intron NP_001306221.1
NM_152640.4 1957 Missense Mutation ACC,TCC T578S NP_689853.3
XM_011520927.2 1957 Missense Mutation ACC,TCC T476S XP_011519229.1

View Full Product Details