Product Details

SNP ID
rs12027398
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:196956167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGAATGGTGTGAGTGGAGACAC[A/T]AAGCCTAACTATATCAGGTGGCAAG
Phenotype
MIM: 600889
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFHR2 PubMed Links

Gene Details

Gene
CFHR2
Gene Name
complement factor H related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312672.1 Intron NP_001299601.1
NM_005666.3 Intron NP_005657.1
XM_005245113.3 Intron XP_005245170.1
XM_011509458.2 Intron XP_011507760.1
XM_011509459.2 Intron XP_011507761.1
XM_011509460.2 Intron XP_011507762.1
XM_017001109.1 Intron XP_016856598.1

View Full Product Details