Product Details
- SNP ID
-
rs12407854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:209432278 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGGCAGCCACCGCCACCACCGCC[A/G]CCGCCACCACCGTAGCAGCAGCAGC
- Phenotype
-
MIM: 613147
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR205
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs565985624] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR205
- Gene Name
- microRNA 205
There are no transcripts associated with this gene.
- Gene
- MIR205HG
- Gene Name
- MIR205 host gene
View Full Product Details