Product Details

SNP ID
rs10917134
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:22057312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCTTCCCCGCCATCTTTGAGAA[A/G]CTGTGTTTCTCTCAAACATAAACAT
Phenotype
MIM: 116952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42 PubMed Links

Gene Details

Gene
CDC42
Gene Name
cell division cycle 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039802.1 Intron NP_001034891.1
NM_001791.3 Intron NP_001782.1
NM_044472.2 Intron NP_426359.1

View Full Product Details