Product Details

SNP ID
rs11171780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56226200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATTCTGTATGGTTTATTCTGA[A/G]GTTCCTAACTTCAGTGAGCCAAACC
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NABP2 PubMed Links

Gene Details

Gene
NABP2
Gene Name
nucleic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024068.3 456 Silent Mutation GAA,GAG E104E NP_076973.1
XM_005269146.2 456 Silent Mutation GAA,GAG E147E XP_005269203.1
XM_005269147.2 456 Silent Mutation GAA,GAG E120E XP_005269204.1
XM_005269148.4 456 Silent Mutation GAA,GAG E104E XP_005269205.1
XM_005269149.4 456 Silent Mutation GAA,GAG E104E XP_005269206.1
Gene
RNF41
Gene Name
ring finger protein 41
There are no transcripts associated with this gene.

Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
There are no transcripts associated with this gene.

View Full Product Details