Product Details

SNP ID
rs13206927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31529265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAAAAAAACCCTCAATCCCACC[A/C]TGCACCACCCCACACCATGCCTACC
Phenotype
MIM: 142560 MIM: 609624
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1G2-DDX39B PubMed Links

Gene Details

Gene
ATP6V1G2-DDX39B
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
DDX39B
Gene Name
DEAD-box helicase 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004640.6 Intron NP_004631.1
NM_080598.5 Intron NP_542165.1
Gene
MCCD1
Gene Name
mitochondrial coiled-coil domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011700.2 Intron NP_001011700.2
Gene
SNORD117
Gene Name
small nucleolar RNA, C/D box 117
There are no transcripts associated with this gene.

View Full Product Details