Product Details
- SNP ID
-
rs12141681
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:54173211 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTATTTATTAAATGTGACACAGC[A/G]GAGAGATTTGTTGCCATTTTTTTTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYB5RL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76755640] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYB5RL
- Gene Name
- cytochrome b5 reductase like
View Full Product Details