Product Details

SNP ID
rs12036530
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:54010650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCCTTCCTCAGTTCCTCCCCAGC[C/T]CTGGTGTGTTCCTGCCCCACACCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDLRAD1 PubMed Links

Gene Details

Gene
LDLRAD1
Gene Name
low density lipoprotein receptor class A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010978.3 Intron NP_001010978.2
NM_001276392.1 Intron NP_001263321.1
NM_001276393.1 Intron NP_001263322.1
NM_001276394.1 Intron NP_001263323.1
NM_001276395.1 Intron NP_001263324.1
XM_011541446.1 Intron XP_011539748.1
XM_017001264.1 Intron XP_016856753.1

View Full Product Details