Product Details

SNP ID
rs12026856
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:53952158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGTTCCCCAAAGGCTTTTCTGT[C/G]GAGCTTTGCATGAACAGGGAAGACG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSPB11 PubMed Links

Gene Details

Gene
HSPB11
Gene Name
heat shock protein family B (small) member 11
There are no transcripts associated with this gene.

Gene
LRRC42
Gene Name
leucine rich repeat containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256409.1 253 Silent Mutation GTC,GTG V53V NP_001243338.1
NM_052940.4 253 Silent Mutation GTC,GTG V53V NP_443172.1
XM_006710327.3 253 Silent Mutation GTC,GTG V53V XP_006710390.1
XM_006710328.3 253 Silent Mutation GTC,GTG V53V XP_006710391.1

View Full Product Details