Product Details

SNP ID
rs12564287
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46610837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGACTATGATAAAAGTTACGAC[A/C]TAAACTCTCCCCAGAAATATGTGCA
Phenotype
MIM: 606724
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MKNK1 PubMed Links

Gene Details

Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
There are no transcripts associated with this gene.

Gene
MOB3C
Gene Name
MOB kinase activator 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145279.4 Intron NP_660322.2
NM_201403.2 Intron NP_958805.1
XM_017000407.1 Intron XP_016855896.1

View Full Product Details