Product Details

SNP ID
rs12732574
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:28747821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCAGGTTACTTTTTAGTTTACT[A/C]TCTTGTAAGAACTGCTGGCACTGGC
Phenotype
MIM: 610640
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
YTHDF2 PubMed Links

Gene Details

Gene
YTHDF2
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172828.1 Intron NP_001166299.1
NM_001173128.1 Intron NP_001166599.1
NM_016258.2 Intron NP_057342.2

View Full Product Details