Product Details

SNP ID
rs12747419
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:28743952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTGATGACTTCTCACACTATG[A/G]GAAACGCCAAGAGGAAGAAGAAAGT
Phenotype
MIM: 610640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YTHDF2 PubMed Links

Gene Details

Gene
YTHDF2
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172828.1 2179 Missense Mutation GAG,GGG E511G NP_001166299.1
NM_001173128.1 2179 Missense Mutation GAG,GGG E561G NP_001166599.1
NM_016258.2 2179 Missense Mutation GAG,GGG E561G NP_057342.2

View Full Product Details