Product Details
- SNP ID
-
hCV31609154
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42104849 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGGACAGACACAAAGAGACAGA[G/T]AGATAAAGAGAGAGACAGAAAGGCA
- Phenotype
-
MIM: 613859
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UMODL1
PubMed Links
Gene Details
- Gene
- UMODL1
- Gene Name
- uromodulin like 1
There are no transcripts associated with this gene.
- Gene
- UMODL1-AS1
- Gene Name
- UMODL1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details