Product Details

SNP ID
rs10775662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39384749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTATCCTCCACCCTCCAAAATA[A/G]AAAAATTTGTTTTTAGAGGCCGGGT
Phenotype
MIM: 602915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WRB PubMed Links

Gene Details

Gene
WRB
Gene Name
tryptophan rich basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146218.1 Intron NP_001139690.1
NM_004627.4 Intron NP_004618.2
Gene
WRB-SH3BGR
Gene Name
WRB-SH3BGR readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317744.1 Intron NP_001304673.1

View Full Product Details