Product Details

SNP ID
rs12133418
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:159284669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCATTAACATTTTGATGAATAG[G/T]TTTCTAATCTTTTATGCATTCATAT
Phenotype
MIM: 147140
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCER1A PubMed Links

Gene Details

Gene
FCER1A
Gene Name
Fc fragment of IgE receptor Ia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002001.3 Intron NP_001992.1

View Full Product Details