Product Details

SNP ID
rs12027623
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:153360394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGGGCAAGACCCTGGAACTCA[A/G]CTTCCTCTTCTATAAATAGAGAATC
Phenotype
MIM: 123886
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
S100A9 PubMed Links

Gene Details

Gene
S100A9
Gene Name
S100 calcium binding protein A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002965.3 Intron NP_002956.1

View Full Product Details