Product Details

SNP ID
rs12445076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15598793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCGTCATTTACTATATCAGTAT[C/T]TCATCGTGGTTTTGTTTTAAACCCC
Phenotype
MIM: 614593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf45 PubMed Links

Gene Details

Gene
C16orf45
Gene Name
chromosome 16 open reading frame 45
There are no transcripts associated with this gene.

Gene
KIAA0430
Gene Name
KIAA0430
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184998.1 Intron NP_001171927.1
NM_001184999.1 Intron NP_001171928.1
NM_014647.3 Intron NP_055462.2
XM_005255704.1 Intron XP_005255761.1
XM_005255706.1 Intron XP_005255763.1
XM_005255707.1 Intron XP_005255764.1
XM_017023900.1 Intron XP_016879389.1
XM_017023901.1 Intron XP_016879390.1
XM_017023902.1 Intron XP_016879391.1
XM_017023903.1 Intron XP_016879392.1
XM_017023904.1 Intron XP_016879393.1
XM_017023905.1 Intron XP_016879394.1
XM_017023906.1 Intron XP_016879395.1

View Full Product Details