Product Details

SNP ID
rs12933123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15402598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAACTCTTGTGATTTCTCTTA[C/G]GGCAGAAATTGAGGACCCCATGGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPV17L PubMed Links

Gene Details

Gene
MPV17L
Gene Name
MPV17 mitochondrial inner membrane protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128423.1 Intron NP_001121895.1
NM_173803.3 Intron NP_776164.2
XM_017023108.1 Intron XP_016878597.1

View Full Product Details