Product Details

SNP ID
rs6861033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34917741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTAGCCATTTAGGAGTTTTGCAG[C/T]TAATATAAGACTCTAAGTAAGTGCA
Phenotype
MIM: 603153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRIX1 PubMed Links

Gene Details

Gene
BRIX1
Gene Name
BRX1, biogenesis of ribosomes
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018321.3 Intron NP_060791.3
Gene
RAD1
Gene Name
RAD1 checkpoint DNA exonuclease
There are no transcripts associated with this gene.

View Full Product Details